L(-)-Glutathion (reduzierte Form)
Lieferant: Chemicon
Synonyms:
L(-)-Glutathion, reduziert
, γ-L-Glutamyl-L-cysteinylglycin
, GSH
CHMCMAB5310EA
1560
CHF
CHMCMAB5310
L(-)-Glutathion (reduzierte Form)
L(-)-Glutathion (reduzierte Form)
Formel:
C₁₀H₁₇N₃O₆S MW: 307,33 g/mol Melting Pt: 185…195 °C Dichte: 1,475 g/cm³ (20 °C) Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00065939 CAS-Nummer: 70-18-8 EINECS: 200-725-4 Merck Index: 13,04488 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...