N-Methylphthalimid ≥98%

Lieferant: Thermo Scientific

Synonyms: N-MethylphthalsäureimidN-Methyl-1,2-benzoldicarbonsäureimid

L08991.14EA 137 CHF
L08991.14
N-Methylphthalimid ≥98%
N-Methylphthalimid
Formel: C₉H₇NO₂
MW: 161,16 g/mol
Siedepunkt: 286 °C (1013 hPa)
Melting Pt: 132…136 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00023063
CAS-Nummer: 550-44-7
EINECS: 208-982-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR