Armadillo™ Low-Profile PCR Strip Plate, 96 Well, Clear
Lieferant: Thermo Scientific
Armadillo™
ADVAAB-2696EA 191 CHF
ADVAAB-2696
Armadillo™ Low-Profile PCR Strip Plate, 96 Well, Clear
Mikrotiterplatten PCR-Platten
Armadillo 96-well low-profile PCR strip plates are the ultimate in high-performance PCR plastics.
  • Black alpha-numeric lettering for easy well identification
  • Integrated frame and well design for improved heat sealing
  • Thin-walled U-bottom wells for optimal heat transfer and maximum sample recovery
Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, Armadillo PCR strip plates provide superior thermal cycling performance under all conditions without warping. The plates can be easily and safely broken into eight-tube strips to help minimise waste.
Low profile design for maximum heat transfer and minimal evaporation with low-volume samples (200 μl working volume). Thermostable polycarbonate semi-skirted frame design for warp-resistance during thermal cycling and enhanced mechanical stability during robotic handling. Optically clear skirt aids in visualisation of liquids and improves stacking capabilities in automated handling systems.