meso-2,3-Dimercaptobernsteinsäure

Lieferant: Molekula

Synonyms: meso-2,3-Dimercapto-1,4-butandisäure(R,S)-2,3-Dimercapto-1,4-butandisäure(R,S)-2,3-DimercaptobernsteinsäureSuccimer

MOLE47679898-5GEA 0 CHF
MOLE47679898-5G
meso-2,3-Dimercaptobernsteinsäure
meso-2,3-Dimercaptobernsteinsäure
Formel: C₄H₆O₄S₂
MW: 182,22 g/mol
Melting Pt: 210…211 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00064799
CAS-Nummer: 304-55-2
EINECS: 206-155-2
Merck Index: 12,09034

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR