Saponin

Lieferant: Roth Carl

ROTH9622.1EA 141.7 CHF
ROTH9622.1
Saponin
Saponin
Dichte: 1,015…1.02 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00081981
CAS-Nummer: 8047-15-2
EINECS: 232-462-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR