Protonex™ Green 500, 1 mg
Lieferant: AAT Bioquest
Protonex™
AATB21215EA 295 CHF
AATB21215
Protonex™ Green 500, 1 mg
Färbemittel Mikrobiologie-Färbemittel
Protonex™ Green dye demonstrated pH-dependent fluorescence.
  • pH-dependent fluorescence
  • No fluorescence outside the cell
  • Excellent for imaging acidic organelles
  • Cell impermeable
Bitte beachten For research use only.