Protonex™ Green 500WS
Lieferant: AAT BIOQUEST
21218
AATB21218EA
307
CHF
AATB21218
Protonex™ Green 500WS
Protonex™ Green 500WS
Protonex™ Green dye demonstrated pH-dependent fluorescence.
- pH-dependent fluorescence
- No fluorescence outside the cell
- Excellent for imaging acidic organelles
- Cell impermeable
Bitte beachten For research use only.