Cl-NERF, 1 mg
Lieferant: AAT Bioquest
AATB21220EA 295 CHF
AATB21220
Cl-NERF, 1 mg
Färbemittel Mikrobiologie-Färbemittel
Cl-NERF is a rhodol dye with pKa = 3,8.
  • pH-dependent fluorescence
  • Ratiometric detection
  • Cell impermeable
Bitte beachten For research use only.