Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Lieferant: AAT Bioquest
Bucculite™
AATB22328EA 683 CHF
AATB22328
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Tests Zelltests
Provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay
  • alternative to anti-BrdU
  • extremely sensitive
  • deep red fluorescence
The resulted iFluor® 647-labeled DNA formed in cells is imaged with a Cy5 filter set.
Uses XdU which is incorporated into cellular DNA during DNA synthesis