Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy7®)
Lieferant: AAT Bioquest
AATB16872EA 221 CHF
AATB16872
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy7®)
Antikörper
AAT Bioquest's anti-rabbit secondary antibodies are affinity-purified antibodies with well-characterised specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target.
  • Affinity-purified
  • Bright signal
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: Cy7®
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit