5(6)-TAMRA Gemisch aus Isomeren ≥85%

Lieferant: AAT BIOQUEST INC

AATB412EA 295 CHF
AATB412
5(6)-TAMRA Gemisch aus Isomeren ≥85%
5(6)-TAMRA

Tetramethylrhodamine-5-(and-6)-maleimide is an excellent reagent for preparing orange fluorescent bioconjugates through thiol modifications of peptides, nucleotides, nucleic acids and proteins.


  • Multiple Bioconjugation Applications
  • 85% Purity


Bitte beachten For research use only.

Formel: C₂₅H₂₂N₂O₅
MW: 430,45 g/mol
MDL Number: MFCD16661194
CAS-Nummer: 98181-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR