Fluo-8FF™ AM
Lieferant: AAT BIOQUEST
Achtung
21104 21105
AATB21104EA 307 CHF
AATB21104 AATB21105
Fluo-8FF™ AM
Fluo-8FF™ AM
Calcium measurements are critical for numerous biological investigations.
  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Suitable for all fluorescence platforms
Bitte beachten For research use only.
MW: 1082,91 g/mol
ADR: 5,II