O15-Palmitoylretinol

Lieferant: CAMBRIDGE ISOTOPE LABS., INC.
Gefahr

Synonyms: Retinol palmitateRetinyl palmitatealle-trans-Retinylpalmitat

CAMBDLM-9309-PKEA 0 CHF
CAMBDLM-9309-PK
O15-Palmitoylretinol
Vitamin A palmitate
Formel: C₃₆H₆₀O₂
MW: 524,87 g/mol
Siedepunkt: 100 °C (212 °F)
Melting Pt: 26 °C (79 °F)
MDL Number: MFCD00019414
CAS-Nummer: 79-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR