γ-Muricholic acid [2,2,3,4,4-D₅] Neat ≥98%, fest

Lieferant: CAMBRIDGE ISOTOPE LABS., INC.
Achtung

CAMBDLM-10628-0.00EA 0 CHF
CAMBDLM-10628-0.00
γ-Muricholic acid [2,2,3,4,4-D₅] Neat ≥98%, fest
γ-Muricholic acid [2,2,3,4,4-D₅]

Bitte beachten Harmful if swallowed. Very toxic to aquatic life with long lasting effects.

Formel: C₂₄D₅H₃₅O₅
MW: 413,6
Melting Pt: 182 °C
Lager Temperatur: Kühlschrank
UN: 3077

Order Now

Spezifikation Testergebnisse

Purity 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR