Powered Air Purifying Respirator System with Speedglas™ 9100 MP Welding Helmet
Lieferant: 3M
Adflo™
MMMA7100054792EA 4270 CHF
MMMA7100054792
Powered Air Purifying Respirator System with Speedglas™ 9100 MP Welding Helmet
Atemschutzmasken Atemschutz-Systeme
The Adflo™ Powered air purifying respirator system with 3M™ Speedglas™ 9100 MP Welding helmet is designed to help protect the eyes and face from radiation, sparks, and spatter. This system also includes respiratory protection, head protection, optional hearing protection, and a flip-up welding shield.
  • Wide, clear visor plate
  • 73×107 mm viewing area
  • Sold with options of 3M™ Speedglas™ 9100 Welding filters
  • Ergonomic, adjustable headband for good comfort, helmet stability, and weight distribution
  • Recommended for construction, heavy infrastructure, and industrial maintenance industries
The flip-up functionality built into the 3M™ Speedglas™ 9100 MP Welding helmet with 3M™ Adflo™ Powered air purifying respirator system makes it an adaptable protection solution. This system allows for quick swapping between welding filter and clear view, helping the user inspect their work while they go.
Recommended applications include metal repair, metal processing, grinding, and welding.
Zertifizierungen: The product meet the requirements of the european community directive 89/686/EEC (personal protective equipment sirective). The product comply with the harmonised european standards EN 175, EN 166 and EN 397. certification under article 10, EC type-examination has been issued by INSPEC International Ltd (notified body number 0194). The complete 3M™ Speedglas™ 9100 MP respiratory systems complies with the harmonised standards EN 12941 and EN 14594. Speedglas 9100 MP welding helmet with powered air respirators or supplied air regulators units marked CE0194 have had certification under article 10, EC-type examination and article 11, EC quality control issued by INSPEC international Ltd. Speedglas 9100 MP welding helmet with powered air respirators or supplied air regulators units marked CE0086 have had certification under article 10, EC-type examination and article 11, EC quality control issued by BSI.
Bestellinformation: Respirator included.