ePrep µSPEed® Cartridge Rack
Lieferant: ePrep
µSPEed®
EPRE01-04161EA 2190 CHF
EPRE01-04161
ePrep µSPEed® Cartridge Rack
Filter Filterkartuschen
The µSPEed Cartridge rack is available where the use of µSPEed cartridge is required. The rack can hold up to 96 cartridges and can be fully integrated with ePrep Workflow.
Bestellinformation: Cartridges sold separately.