Superblast Safety Goggles
Lieferant: Bollé Safety
BOLESUPBLFAPSIEA 40 CHF
BOLESUPBLFAPSI BOLESUPBLAPSIP BOLESUPBLEPSI BOLESUPBLAWPCC5
Superblast Safety Goggles
Vollsichtbrillen
Superblast goggles gets softer frame, a platinum version and a simplified intuitive assembly system goggle-Visor.
  • Flexible and clear frame for increased comfort
  • Wide choice of frame types for all applications
  • Intuitive and simple goggle-visor assembly system
Lens: Polycarbonate (2 mm)
Curve: Slightly curved
Frame material: PVC
Strap: Nylon Polyster (250 mm)