Hermetic Plastic Containers
Lieferant: Multiroir
MULC130278EA 64.7 CHF
MULC130278
Hermetic Plastic Containers
Probenbehälter Universalbehälter
Hermetic plastic containers.
  • Polypropylene construction
  • Reinforced lid
  • Stackable and very stable when stacked
  • Lightweight and strong at the same time