Safety Storage Cabinet, Type 90, ION-CORE-90 CHARGE model IO90.195.060.CC.WDC
Lieferant: asecos
MSGA41374EA 2640 CHF
MSGA41371 MSGA41373 MSGA41374 MSGA41375 135-2885 MSGA41376 MSGA41366 MSGA41369 MSGA41370
Safety Storage Cabinet, Type 90, ION-CORE-90 CHARGE model IO90.195.060.CC.WDC
Möbel Lagerschränke
Safe active and passive storage of lithium-ion batteries.
  • Robust construction and longevity
  • Integrated potential-free alarm contact
  • No unauthorised use
  • Integrated transport base
  • Easy alignment
  • Smoke detector
  • Pressure relief
  • All-around protection
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface. Integrated potential-free alarm contact for strongly recommended alarm forwarding. Optionally available with module for remote signalling.
No unauthorised use: door lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green). Permanent locking of the door by automatic 2-point locking. Integrated transport base: In the event of an emergency, immediate evacuation and transport of the cabinet to a safe place outdoors.
Easy alignment: adjusting aids to compensate for uneven floor. Smoke detector for fire detection and forwarding the alarm to the building management system. Pressure relief in the head section.
All-around protection: 90 minutes fire protection from outside to inside (type 90/type tested in accordance with EN 14470-1) and for more than 90 minutes fire resistance for fires from inside to outside.
Zertifizierungen: Fire resistance 90 minutes. Type tested in accordance with EN 14470-1. CE compliant.
Bestellinformation: Interior equipment packages need to be ordered separately (not included). Please contact your Avantor representative for more information and available options.
Lieferumfang: Delivery prices available upon request.
Bitte beachten According to the security concept it is highly recommended to place ION-LINE cabinets at the ground level in a location from where a quick evacuation from the building is possible in the event of fire.