Tanker Spill Response Kits
Lieferant: 3M
TSK15 TSK30N
MMMATSK15EA 150 CHF
MMMATSK15 MMMATSK30N
Tanker Spill Response Kits
Absorptionsmaterial
3M™ Tanker spill response kit is the modern and effective way to control and clean up spills, particularly those resulting from liquid fuel deliveries. The kit is easily stowed in confined areas and is readily available for rapid deployment.
  • Designed to absorb and contain small spills and leaks
  • Compact size convenient for trucks, shipping departments, and for areas of oil transfer
  • Easily stowed in confined areas, such as behind the drivers seat
Kits contains 50 sheets, 2 mini-booms, 1 disposal bag and tie, and 1 disposal guide and label. Absorbency per kit: 30 litres (TSK-30) or 15 litres (TSK-15).