Liquid Weighing Pour Boats, Polystyrene, White
Lieferant: Heathrow Scientific
611-5278EA 83.1 CHF
611-5278 611-5277
Liquid Weighing Pour Boats, Polystyrene, White
Schiffchen Wägeschiffchen
Convenient and specific tool for customers who need to weigh liquids on a scale. Deep vessel with sturdy handle designed for convenient handling and pouring of liquids.
  • Flat bottom ensures stability and prevents tipping
  • Notched pour spout design controls liquid pour
  • Operating temperature range 0 to 40 °C (104 °F)
  • Made from FDA food grade polystyrene in an ISO Class 8 clean room
  • Material biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
  • Recyclable, marked
Available in 3 sizes (50, 100 and 200 ml).