TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether) -Lösung 70% in Wasser ≥70%
Lieferant: Spectrum Chemical
Gefahr
TR140-500ML TR140-4LT TR140-20LTMT
SPCMTR140-500MLEA 195 CHF
SPCMTR140-500ML SPCMTR140-4LT SPCMTR140-20LTMT
TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether) -Lösung 70% in Wasser ≥70%
TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether)
Triton(R) X-405, 70 Percent Solution in Water is used as an excellent emulsion stabiliser that provides freeze/thaw and ionic stability.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel: C₂₈H₅₀O₈
MW: 514.69 g/mol
Siedepunkt: 120
Melting Pt: -4
Dichte: 1,096
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01779734
CAS-Nummer: 9002-93-1
Merck Index: 12,06858
Order Now
Spezifikation Testergebnisse
ACTIVE CONTENT 69-71%
pH OF A 1% SOLUTION IN 62.5% IPA @ 25°C 7.0-9.0
COLOR (APHA) max 250