Sodium fluoride ≥98,5%, Pulver USP, Ph. Eur., BP
Lieferant: Spectrum Chemical
|
|
SPCMSO167-125GMEA
204
CHF
SPCMSO167-125GM
Sodium fluoride ≥98,5%, Pulver USP, Ph. Eur., BP
Sodium fluoride
Sodium Fluoride, Powder, USP, EP, BP is used to prevent cavities making teeth stronger and more resistant to decay caused by acid and bacteria. Toothpaste being the preferred delivery method. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Skull and crossbones;Exclamation mark;Health hazard
UN1690
Formel:
NaF MW: 41,99 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003524 CAS-Nummer: 7681-49-4 UN: 1690 ADR: 6.1,III |
Spezifikation Testergebnisse
Assay (NaF; Dried Basis) | 98.5 - 100.5% |
Acidity or Alkalinity | To Pass Test |
Appearance of Solution | To Pass Test |
Loss On Drying | max. 0.5% |
Fluosilicate | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...