Potassium bromide ≥98,0% USP
Lieferant: Spectrum Chemical
|
|
Synonyms:
Bromkalium
SPCMP1742-25KGEA
33680
CHF
SPCMP1742-25KG
SPCMP1742-2.5KG
Potassium bromide ≥98,0% USP
Kaliumbromid
Potassium Bromide, USP is used in veterinary medicine to treat epilepsy in dogs.
All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
KBr MW: 119 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00011358 CAS-Nummer: 7758-02-3 |
Spezifikation Testergebnisse
Assay (Dried Basis) | 98.0 - 100.5% |
Acidity or Alkalinity | To Pass Test |
Loss On Drying | max. 1.0 % |
Chlorine | max. 0.6 % |
Sulfates | max. 0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...