For the measurement of anti-ovalbumin of the IgG1 isotype in mouse plasma and serum.
- Measure mouse anti-OVA IgG1 in mouse plasma, serum, or BAL fluids
- Employs an affinity-purified mouse anti-OVA IgG1 polyclonal antibody standard for accurate results
- Assay Range: 1,56 to 200 ng/ml with an LLOQ of 1,56 ng/ml
- Rapid assay, get results in under four hours
Cayman's ovalbumin IgG1 (mouse) ELISA kit is an immunometric (i.e., 'sandwich') assay which can be used to measure anti-ovalbumin of the IgG₁ isotype in mouse plasma and serum without prior sample purification.
Lieferumfang: Contents: antibody, standard, buffer, polysorbate 20, substrate solution, stop solution, plate, and cover sheet