Soluble Collagen Type I Solution, Bovine
Lieferant: Viscofan Bioengineering
MSPP500060635EA 0 CHF
MSPP500060635
Soluble Collagen Type I Solution, Bovine
Tests Proteintests
Soluble bovine type I collagen acid extracted (Telocollagen) at 5 mg/ml. Soluble collagen can be used as a thin coating for 2D cell culture in any cell culture dish. For use in 3D cell culture, collagen hydrogels, and bioinks, sterile 20 ml bottle.
  • Highly purified collagen type I
  • Produced without enzymatic treatment
  • Biocompatible
  • Uses: 2D and 3D cell culture, tissue engineering, hydrogels, bioprinting
Soluble collagen is derived from highly purified bovine type I collagen and provides in-vivo-like conditions to positively influence adherence, growth and differentiation of your cells. The nature-like matrix promotes authentic cell performance and enables novel developments in: Tissue engineering e.g. of skin, osteogenic, endothelial, hepatocytic and other tissues, analysis of cell migration, e.g. at metastasis, wound healing or development, thin coating for 2D cell culture in any cell culture dish and 3D cell culture and bioprinting and collagen hydrogels.
In addition, this flexible matrix may be mixed with factors like other ECM components, antibiotics or growth factors, e.g. for directed cell manipulation. Supplemental cross-linking by the user is also feasible. The product is supplied as an aqueous solution (0,5%) in 0,1 M acetic acid with a protein concentration of approximately 5,0 mg/ml. Collagen Type I solution is sterile-filtered and is supplied as a ready-to-use solution, providing the flexibility to dilute to lower concentrations.
Zertifizierungen: ISO 9001.