Argos Technologies PolarSafe® Upright Freezer Racks
Lieferant: Argos Technologies
04404-03 04398-26 04406-45 04406-47
ARGT04404-03EA 0 CHF
ARGT04404-03 ARGT04398-26 ARGT04406-45 ARGT04406-47
Argos Technologies PolarSafe® Upright Freezer Racks
Gestelle Kryogestelle
Get organised for easier retrieval of samples.
  • Sturdy corrosion-resistant stainless steel construction for long-term use
  • Convenient handles make sample retrieval easy
Top loading sliding tray eco-racks are a great economical alternative to the standing side access drawer racks. Trays slide out for easy retrieval of samples without having to remove the entire rack from the freezer.
Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer.
Upright freezer racks for 0,2 ml tube 96-well PCR boxes feature handles at both ends to accommodate freezers with right- or left-handed doors.