Axygen® Robotic Tips for Corbett X-tractor
Lieferant: Corning
Axygen®
736-0522EA 632 CHF
736-0522
Axygen® Robotic Tips for Corbett X-tractor
Pipettenspitzen
Tips in 96-well format designed for Corbett X-tractor Gene 8 channel head.
  • Clear polypropylene
  • With aerosol-resistant filters
  • Optimised fine-point for accurate dispensing
  • Certified RNase, DNase and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.