ADCF Antifoam agent, irradiated , HyClone
Lieferant: HyClone products (Cytiva)
HYCLSH30897.02EA 0 CHF
HYCLSH30897.02
ADCF Antifoam agent, irradiated , HyClone
Zellkultur-Ergänzungen und -Zusätze
Prevents and minimizes foaming in bioprocess cell culture systems.
  • Irradiated Antifoam effectively prevents and dissipates foaming in cell culture systems.
  • Provided in easy-to-use single-use bags.
  • Designed with tubing lines and fittings to enable easy connection and delivery to any cell culture system (customization possible).
  • HyClone™ Antifoam is an emulsion with ~3% active simethicone.