3,3,3-Trifluoro-2-oxopropanoic acid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Trifluoropyruvic acid, 3,3,3-Trifluoropyruvic acid

AS28102G5 AS28102G1
ACTIAS28102G5EA 584 CHF
ACTIAS28102G5 ACTIAS28102G1
3,3,3-Trifluoro-2-oxopropanoic acid
3,3,3-Trifluoro-2-oxopropanoic acid
Formel: C₃HF₃O₃
MW: 142,03 g/mol
MDL Number: MFCD06203569
CAS-Nummer: 431-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR