Propafenone Related Compounds, Reference Standard
Lieferant: USP
1570304 1570326 1570315
1570304.EA 443 CHF
1570304. 1570326. 1570315.
Propafenone Related Compounds, Reference Standard
Standards Organische Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Zertifizierungen: US Pharmacopea Reference Standard
Order Now
TECHNISCHE DATEN
Bezeichnung Propafenone Hydrochloride Propafenone Related Compound B Propafenone Related Compound A
Synonym(s) RS,E)-1-[2-[2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylprop-2-en-1-one N-{2-Hydroxy-3-[2-(3-phenylpropanoyl)phenoxy]propyl}-N-propylformamide
Environmentally Preferable
CAS Nr. 34183-22-7 88308-22-9
Formel C₂₁H₂₈ClNO₃ C₂₁H₂₅NO₃