SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
Lieferant: G Bioscienes
Synonyms: SMCC
BC20EA 615 CHF
BC20
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
Succinimidyl 4-(N-maleimidomethyl) cyclohexane-1-carboxylate Heterobifunctional Cross Linkers.
  • Ideal for enzyme-antibody crosslinking
Formel: C₁₆H₁₈N₂O₆
MW: 334,33 g/mol
MDL Number: MFCD00009634
CAS-Nummer: 64987-85-5
Order Now
Spezifikation Testergebnisse
Spacer Arm (Å) 11.6
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable YES
Water Soluble NO
Cleavable/ Reversible NO