Anti- NEFL Rabbit polyclonal antibody

Lieferant: Novus Biologicals
NOVUNB300-131EA 635 CHF
NOVUNB300-131
Anti- NEFL Rabbit polyclonal antibody
Antikörper
The NF-L Antibody from Novus Biologicals is a rabbit polyclonal antibody to NF-L. This antibody reacts with human, mouse, rat, porcine, avian, bovine, mammal. The NF-L Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:5000

Type: Primary
Antigen: NF-L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Avian, Bovine, Mammal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR