Adjustable Mechanical Feed Peristaltic Pumps
Lieferant: Stenner
MFLX74206-15EA 757 CHF
MFLX74206-15 MFLX74207-00 MFLX74206-45 MFLX74206-00 MFLX74206-10 MFLX74206-20 MFLX74207-05 MFLX74206-30 MFLX74206-40
Adjustable Mechanical Feed Peristaltic Pumps
Pumpen Peristaltikpumpen
Exceptional reliability and ease of service reduce maintenance and cost.
  • Polycarbonate corrosion-resistant housing
  • 24/7 continuous duty cycle
  • Mount pumps vertically or horizontally
Single Head, Adjustable Mechanical Feed Rate Pumps run reliably and consistently against 100 psi of pressure. These adjustable models feature a 20:1 turndown ratio with an adjustable rate in 2.5% increments from 5 to 100%.