Granular Activated Carbon Cartridges for "Big Blue" housing
Lieferant: Big Blue
MFLX01512-66EA 119 CHF
MFLX01512-66 MFLX01512-64
Granular Activated Carbon Cartridges for "Big Blue" housing
Filter Filterkartuschen
Remove organic tastes/odors and chlorine.
  • Constructed with granular activated carbon (GAC)
These cartridges remove organic tastes/odors and chlorine. Special design maximizes contact time and reduces channeling or bypass that is common with GAC cartridges. Included within the cartridge is a 20-micron post-filter to reduce carbon fines††.
The 20" cartridge meets NSF Standard 42 for materials.