Micro volume Pipette tips, MµltiFit™
Lieferant: Sorenson Bioscience
37650 13980
SORE37650EA 63.6 CHF
SORE37650 SORE13980
Micro volume Pipette tips, MµltiFit™
Pipettenspitzen
These natural-colored tips fit most popular brand research-grade pipettors.
They feature a narrow bore that reduces dead air space around extended air-displacement piston. Narrow bore also increases accuracy and reproducibility for critical ultra microvolume ranges.
Zertifizierungen: Certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Bestellinformation: Available in autoclavable polypropylene racks, bulk packed, or QuickRack reloads for the reloading racks.
Verpackung: Contents: Casein peptone (10 g/L), UF Yeast Powder (5 g/L), NaCl (10 g/L)