Mark 7 spray chamber supplies
Lieferant: VARIAN
7210027600 7210027700 3710009200 810133400 1510225000 1510226000 1510226500 1510226600 1510226700 1510226800 1610118400 310046600 810134300 810134400 6910015200
VARI7210027600EA 114 CHF
VARI7210027600 VARI7210027700 HEWL3710009200 HEWL810133400 HEWL1510225000 HEWL1510226000 HEWL1510226500 HEWL1510226600 HEWL1510226700 HEWL1510226800 HEWL1610118400 HEWL310046600 HEWL810134300 HEWL810134400 HEWL6910015200
Mark 7 spray chamber supplies
Chromatographie-Zubehör Chromatographie-Ersatzteile
Mark 7 Spray Chambers for flame atomisation System. Easy-to-use twist and lock assembly allows for easy cleaning and routine maintenance.
  • Inert construction makes spray chamber suitable for use with aqueous and organic solutions
  • Fit organic resistant O-rings to ensure organic compatibility
Flexible operation with the capability to tune performance using the externally adjustable impact bead, and to fit mixing paddles for better precision, or both.
Lieferumfang: Mark 7 operating supplies kit available to support routine operation. Contents: VenturiMore Product Information