Banana powder
Lieferant: Apollo Scientific
APOSBIPB852-100GEA 17.6 CHF
APOSBIPB852-100G
Banana powder
Gastronomiezubehör
A mixture of natural banana puree and maltodextrin. Use at a concentration of 30 to 50 g/L.
  • MFCD00130601