Wipedown dry wipes, VAI®
Lieferant: VELTEK ASSOCIATES
VEL812X12S3002 VEL812X12S3031
VELTVEL812X12S3002EA 1350 CHF
VELTVEL812X12S3002 VELTVEL812X12S3031
Wipedown dry wipes, VAI®
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
WipeDown is designed as a dry cleaning wipe for use during wipe downs of filling and packaging machinery, stainless steel surfaces, lexan, polycarbonate, glass and any other critical surface that requires cleaning.
  • Laundered polyester
  • Laser cut sealed edge
  • Low particulate and low extractable
These dry wipes are constructed of a 100% continuous polyester fiber that is designed to be clean and absorbent.
VAI®'s dry wipes are ultra-clean, superior, laundered dry wipes suitiable for use in a cleanroom environment. Specifically designed to be clean and absorbent, using FocusEdge cutting technology to cut wipes that seal the edges and reduce shedding to a minimum during wipe down or general use.
Avaliable in two sorbancy options. WipeDown Dry Wipes are ideal for use in both aspetic and non-aseptic wipe downs on any critical surface that requires cleaning. WipeDown HighSorb Dry Wipes are ideal for use in both aseptic and non-aseptic areas to keep the surface dry, residue-free, and streak-free after chemical application.
Verpackung: Each sterile bag is bulk packaged in an additional single outer bag pack.