Recombinant TrypZean®
Lieferant: Apollo Scientific
APOSBIT0530-10MGEA 200 CHF
APOSBIT0530-10MG APOSBIT0530-5MG
Recombinant TrypZean®
Enzyme
Plant origin, non-animal source bovine trypsin. Recombinantly expressed in corn, completely free of animal source contaminants. Intended for use in cell culture.