Yeast hexokinase
Lieferant: Apollo Scientific
APOSBIB3075-500UEA 166 CHF
APOSBIB3075-500U
Yeast hexokinase
Enzyme
From Yeast (Saccharomyces cerevisiae). Specific activity: IU/mg protein>180 units. IU per 1ml solution is approximately 1000 units
  • Ammonium sulfate suspension