Photometric tube tests, NANOCOLOR® ortho- and Total Phosphate
Lieferant: MACHEREY-NAGEL
985081 985076 985079 985055 985095 985080
MANA985081EA 158 CHF
MANA985081 MANA985076 MANA985079 MANA985055 MANA985095 MANA985080
Photometric tube tests, NANOCOLOR® ortho- and Total Phosphate
Umwelt-Prüfkits
A pre-dosed NANOCOLOR® tube test for photometric determination of ortho- and total phosphate in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.