Photometric tube tests, NANOCOLOR® cyanide
Lieferant: MACHEREY-NAGEL
985031
985025
MANA985031EA
122
CHF
MANA985031
MANA985025
Photometric tube tests, NANOCOLOR® cyanide
Umwelt-Prüfkits
A pre-dosed NANOCOLOR tube test for photometric determination of cyanide in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.