2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Lieferant: Apollo Scientific
Achtung

Synonyms: 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzonitrile, 4-Cyano-3,5-difluorobenzeneboronic acid pinacol ester

PC408301-5G PC408301-250MG
APOSPC408301-5GEA 0 CHF
APOSPC408301-5G APOSPC408301-250MG
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Formel: C₁₃H₁₄BF₂NO₂
MW: 265,067 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD22419275
CAS-Nummer: 1003298-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR