3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde
Lieferant: Apollo Scientific
PC300649-1G
PC300649-10G
PC300649-5G
APOSPC300649-1GEA
237
CHF
APOSPC300649-1G
APOSPC300649-10G
APOSPC300649-5G
3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde
3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde
Formel:
C₁₂H₇F₃N₂O₂ Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD11505008 CAS-Nummer: 1216892-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...