4-Fluorobenzylmagnesium chloride 0,25 M in THF

Lieferant: Apollo Scientific
Gefahr

APOSPC2833-100MLEA 916 CHF
APOSPC2833-100ML
4-Fluorobenzylmagnesium chloride 0,25 M in THF
4-Fluorobenzylmagnesium chloride
Formel: C₇H₆ClFMg
MW: 168,878 g/mol
Siedepunkt: 65 °C
Dichte: 0,948 g/cm³ (25 °C)
Flash-Pt: <–17 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD01319898
CAS-Nummer: 1643-73-8
UN: 2924
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR