Benzotrifluorid 99%

Lieferant: Apollo Scientific
Gefahr

Synonyms: Lösungsmittel BTFα,α,α-Trifluortoluol

APOSPC1190-1KGEA 180 CHF
APOSPC1190-1KG
Benzotrifluorid 99%
Benzotrifluorid

An alternative solvent to dichloromethane with higher bp and lower toxicity.

Formel: C₇H₅F₃
MW: 146,11 g/mol
Siedepunkt: 101…102 °C
Melting Pt: –29 °C
Dichte: 1,199 g/cm³ (25 °C)
Flash-Pt: 12 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000372
CAS-Nummer: 98-08-8
EINECS: 202-635-0
UN: 2338
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR