3-Chlor-2,5,6-trifluorpyridin

Lieferant: Apollo Scientific
Gefahr

APOSPC10520-25GEA 401 CHF
APOSPC10520-25G
3-Chlor-2,5,6-trifluorpyridin
3-Chloro-2,5,6-trifluoropyridine
Formel: C₅HClF₃N
Siedepunkt: 132…134 °C
Flash-Pt: >100 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03001160
CAS-Nummer: 2879-42-7
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR