Ethyl-2-amino-4-methylthiazol-5-carboxylat 98%

Lieferant: Apollo Scientific
Achtung

APOSOR9899-25GEA 61.2 CHF
APOSOR9899-25G
Ethyl-2-amino-4-methylthiazol-5-carboxylat 98%
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Formel: C₇H₁₀N₂O₂S
MW: 186,22 g/mol
Melting Pt: 172…180 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00123414
CAS-Nummer: 7210-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR