6-Bromoquinazolin-4-amine 98%

Lieferant: Apollo Scientific
Achtung

OR932284-5G
APOSOR932284-5GEA 156 CHF
APOSOR932284-5G
6-Bromoquinazolin-4-amine 98%
6-Bromoquinazolin-4-amine
Formel: C₈H₆BrN₃
MW: 224,0600 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD02228849
CAS-Nummer: 21419-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR